Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sampling Scoop
Synonym(s):
measuring scoop, powder sampler
Products
Product Category
spatulas/scoops (8)
Manufacturer
Capacity (mL)
Length (in)
Color
Package Size
Sterility
Sterilization Method
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
HS120855
Sampling Scoop,
capacity 125 mL, sterile; electron beam irradiated
Expand
View Pricing
HS120854
Sampling Scoop,
capacity 250 mL, non-sterile
Expand
View Pricing
HS120853
Sampling Scoop,
capacity 250 mL, sterile; electron beam irradiated
Expand
View Pricing
HS120857
Sampling Scoop,
capacity 60 mL, sterile; electron beam irradiated
Expand
View Pricing
HS120852
Sampling Scoop,
capacity 500 mL, non-sterile
Expand
View Pricing
HS120851
Sampling Scoop,
capacity 500 mL, sterile; electron beam irradiated
Expand
View Pricing
HS120858
Sampling Scoop,
capacity 60 mL, non-sterile
Expand
View Pricing
HS120856
Sampling Scoop,
capacity 125 mL, non-sterile
Expand
View Pricing