Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Round crystal window
Products
Brand
Sigma-Aldrich (17)
Diameter (mm)
Technique
Filter Products
Showing 1-18 of 18
Compare
Product Number
Description
Pricing
Z123595
Round crystal window,
NaCl, diam. × thickness 25 mm × 4 mm
Expand
View Pricing
Z123609
Round crystal window,
KBr, diam. × thickness 25 mm × 4 mm
Expand
View Pricing
Z267724
Round crystal window,
NaCl, diam. × thickness 25 mm × 2 mm
Expand
View Pricing
Z267627
Round crystal window,
KBr, diam. × thickness 25 mm × 2 mm
Expand
View Pricing
Z267635
Round crystal window,
KBr, diam. × thickness 25 mm × 5 mm
Expand
View Pricing
Z805289
Round crystal window,
KBr, diam. × thickness 20 mm × 2 mm
Expand
View Pricing
Z267562
Round crystal window,
CaF
2
, diam. × thickness 25 mm × 2 mm
Expand
View Pricing
Z805165
Round crystal window,
NaCl, diam. × thickness 22 mm × 4 mm
Expand
View Pricing
Z267694
Round crystal window,
KRS-5, diam. × thickness 25 mm × 4 mm
Expand
View Pricing
Z267813
Round crystal window,
ZnSe, diam. × thickness 25 mm × 4 mm
Expand
View Pricing
Z267732
Round crystal window,
NaCl, diam. × thickness 25 mm × 5 mm
Expand
View Pricing
Z267805
Round crystal window,
ZnSe, diam. × thickness 25 mm × 2 mm
Expand
View Pricing
Z123633
Round crystal window,
ZnS (Irtran-2), diam. × thickness 25 mm × 2 mm
Expand
View Pricing
Z804622
Round crystal window,
CaF
2
, diam. × thickness 13 mm × 2 mm
Expand
View Pricing
Z267686
Round crystal window,
KRS-5, diam. × thickness 25 mm × 2 mm
Expand
View Pricing
Z267791
Round crystal window,
ZnSe, diam. × thickness 13 mm × 2 mm
Expand
View Pricing
Z267643
Round crystal window,
KBr, diam. × thickness 38 mm × 6 mm
Expand
View Pricing
Z267651
Round crystal window,
KBr, diam. × thickness 49 mm × 6 mm
Expand
View Pricing