
Ritanserin

Synonym(s):
6-[2-[4-(bis(4-Fluorophenyl)methylene]-1-piperidinyl]ethyl]-7-methyl-5H-thiazolo[3,2-a]pyrimidin-5-one
Empirical Formula (Hill Notation):
C27H25F2N3OS
CAS Number:
Molecular Weight:
477.57
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing