Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Riboflavin 5′-monophosphate sodium salt hydrate
Synonym(s):
FMN-Na, Flavin mononucleotide, Riboflavin 5′-phosphate sodium salt
Empirical Formula (Hill Notation):
C
17
H
20
N
4
NaO
9
P · xH
2
O
CAS Number:
130-40-5
Molecular Weight:
478.33 (anhydrous basis)
EC Number:
204-988-6
Products
Product Category
nucleotides (3)
Brand
Sigma-Aldrich (3)
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
F2253
Riboflavin 5′-monophosphate sodium salt hydrate,
synthetic, ≥70% (HPLC)
Expand
View Pricing
F6750
Riboflavin 5′-monophosphate sodium salt hydrate,
73-79% (fluorimetric)
Expand
View Pricing
F1392
Riboflavin 5′-monophosphate sodium salt hydrate,
suitable for electrophoresis, suitable for acrylamide photopolymerization, ≥70%, powder
Expand
View Pricing