Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Resin-bonded glass fibre board (thermal conductivity)
Products
Manufacturer
Physical Form
Agency
Format
Application
Reference Material Type
Special Grade
certified reference material (2)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
ERMFC440B
Resin-bonded glass fibre board (thermal conductivity),
ERM
®
, certified reference material, size 50 cm × 50 cm
Expand
View Pricing
ERMFC440C
Resin-bonded glass fibre board (thermal conductivity),
ERM
®
, certified reference material, size 60 cm × 60 cm
Expand
View Pricing
IRMM440C
Resin-bonded glass fibre board (thermal conductivity),
IRMM
®
, certified reference material
Expand
View Pricing
IRMM440A
Resin-bonded glass fibre board (thermal conductivity),
IRMM
®
, certified reference material
Expand
View Pricing