Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Reservoir
Products
Manufacturer
Capacity (mL)
Package Size
Sterility
Filter Products
Showing 1-19 of 19
Compare
Product Number
Description
Pricing
AXYRESV25S
Reservoir,
capacity 25 mL, sterile
Expand
View Pricing
AXYRESV25
Reservoir,
capacity 25 mL
Expand
View Pricing
AXYRESMW12HP
Reservoir,
12 channel, high profile multiwells, size 7 mL, 25 ea of
Expand
View Pricing
AXYRESMW12LP
Reservoir,
12 channel, low profile multiwells, size 1.75 mL, 25 ea of
Expand
View Pricing
AXYRESV25SI
Reservoir,
capacity 25 mL, individually wrapped, sterile
Expand
View Pricing
AXYRESV50
Reservoir,
capacity 50 mL
Expand
View Pricing
AXYRESMW8HP
Reservoir,
8 channel, high profile multiwells, size 10 mL, 25 ea of
Expand
View Pricing
AXYRESMW8LP
Reservoir,
8 channel, low profile multiwells, size 4 mL, 25 ea of
Expand
View Pricing
AXYRESMW4HP
Reservoir,
high profile multi-well, 4 channel
Expand
View Pricing
AXYRESV50S
Reservoir,
capacity 50 mL, sterile
Expand
View Pricing
AXYRESSW384FX
Reservoir,
384 bottom, medium profile single-well
Expand
View Pricing
AXYRESV100S
Reservoir,
capacity 100 mL, sterile
Expand
View Pricing
AXYRES2CV25S
Reservoir,
capacity 25 mL, divided, sterile
Expand
View Pricing
AXYRESV100
Reservoir,
capacity 100 mL
Expand
View Pricing
AXYRES2CV25SI
Reservoir,
capacity 25 mL, divided, individually wrapped, sterile
Expand
View Pricing
AXYRESSW384LP
Reservoir,
384 bottom troughs, low profile single well, size 92 mL, 25 ea of
Expand
View Pricing
AXYRES2CV25
Reservoir,
capacity 25 mL, divided
Expand
View Pricing
AXYRESV100SI
Reservoir,
capacity 100 mL, individually wrapped, sterile
Expand
View Pricing
AXYRESSW384FXSI
Reservoir,
384 bottom, medium profile single-well, individually wrapped, sterile
Expand
View Pricing