Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Refractive Index Standard at 20 °C 25 °C and 30 °C
Products
Manufacturer
Application
Reference Material Type
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
PSRI01
Refractive Index Standard at 20 °C 25 °C and 30 °C,
Nominal RI Value 1.3325 at 25 °C, in 10mL Tamper-Evident Glass Vials
Expand
View Pricing
PSRI02
Refractive Index Standard at 20 °C 25 °C and 30 °C,
Nominal RI Value 1.3891 at 25 °C, in 10mL Tamper-Evident Glass Vials
Expand
View Pricing
PSRI07
Refractive Index Standard at 20 °C 25 °C and 30 °C,
Nominal RI Value 1.4941 at 25 °C, in 10mL Tamper-Evident Glass Vial
Expand
View Pricing
PSRI10
Refractive Index Standard at 20 °C 25 °C and 30 °C,
Nominal RI Value 1.6556 at 25 °C, in 10mL Tamper-Evident Glass Vials
Expand
View Pricing
PSRI08
Refractive Index Standard at 20 °C 25 °C and 30 °C,
Nominal RI Value 1.5349 at 25 °C, in 10mL Tamper-Evident Glass Vials
Expand
View Pricing
PSRI09
Refractive Index Standard at 20 °C 25 °C and 30 °C,
Nominal RI Value 1.5440 at 25 °C, in 10mL Tamper-Evident Glass Vials
Expand
View Pricing
PSRI03
Refractive Index Standard at 20 °C 25 °C and 30 °C,
Nominal RI Value 1.4023 at 25 °C, in 10mL Tamper-Evident Glass Vials
Expand
View Pricing
PSRI06
Refractive Index Standard at 20 °C 25 °C and 30 °C,
Nominal RI Value 1.4573 at 25 °C, in 10mL Tamper-Evident Glass Vials
Expand
View Pricing
PSRI09K
Refractive Index Standard at 20 °C 25 °C and 30 °C,
Nominal RI Value 1.5440 at 25 °C, in 5 x 10mL Tamper-Evident Glass Vials
Expand
View Pricing
PSRI05
Refractive Index Standard at 20 °C 25 °C and 30 °C,
Nominal RI Value 1.4206 at 25 °C, in 10mL Tamper-Evident Glass Vials
Expand
View Pricing
PSRI10K
Refractive Index Standard at 20 °C 25 °C and 30 °C,
Nominal RI Value 1.6556 at 25 °C, in 5 x 10mL Tamper-Evident Glass Vials
Expand
View Pricing
PSRI08K
Refractive Index Standard at 20 °C 25 °C and 30 °C,
Nominal RI Value 1.5349 at 25 °C, in 5 x 10mL Tamper-Evident Glass Vials
Expand
View Pricing
PSRI07K
Refractive Index Standard at 20 °C 25 °C and 30 °C,
Nominal RI Value 1.4941 at 25 °C, in 5 x 10mL Tamper-Evident Glass Vials
Expand
View Pricing
PSRI02K
Refractive Index Standard at 20 °C 25 °C and 30 °C,
Nominal RI Value 1.3891 at 25 °C, in 5 x 10mL Tamper-Evident Glass Vials
Expand
View Pricing
PSRI03K
Refractive Index Standard at 20 °C 25 °C and 30 °C,
Nominal RI Value 1.4023 at 25 °C, in 5 x 10mL Tamper-Evident Glass Vials
Expand
View Pricing
PSRI05K
Refractive Index Standard at 20 °C 25 °C and 30 °C,
Nominal RI Value 1.4206 at 25 °C, in 5 x 10mL Tamper-Evident Glass Vials
Expand
View Pricing