
Ramipril impurity B

Synonym(s):
(2S,3aS,6aS)-1-[(S)-2-[[(S)-1-[(Methylethoxy)carbonyl]-3-phenylpropyl]amino]propanoyl]octahydrocyclopenta[b]pyrrole-2-carboxylic acid, Ramipril isopropyl ester
Empirical Formula (Hill Notation):
C24H34N2O5
CAS Number:
Molecular Weight:
430.54
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing