(R)-(−)-2-Oxothiazolidine-4-carboxylic acid
Synonym(s):
L-(−)-2-Oxothiazolidine-4-carboxylic acid
Empirical Formula (Hill Notation):
C4H5NO3S
CAS Number:
Molecular Weight:
147.15
Products
Product Category
Brand
Color
Formula Weight
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing