Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Quickfit
®
round-bottom flasks
Products
Product Category
flasks (27)
Manufacturer
Capacity (mL)
Closure Type
Package Size
Sterility
Filter Products
Showing 1-20 of 27
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
Z302775
Quickfit
®
round-bottom flasks,
capacity 1,000 mL, joint: ST/NS 24/29
Expand
View Pricing
Z302767
Quickfit
®
round-bottom flasks,
capacity 500 mL, joint: ST/NS 24/29
Expand
View Pricing
Z302821
Quickfit
®
round-bottom flasks,
capacity 250 mL, joint: ST/NS 29/32
Expand
View Pricing
Z302872
Quickfit
®
round-bottom flasks,
capacity 5,000 mL, joint: ST/NS 29/32
Expand
View Pricing
Z302732
Quickfit
®
round-bottom flasks,
capacity 100 mL, joint: ST/NS 24/29
Expand
View Pricing
Z302570
Quickfit
®
round-bottom flasks,
capacity 5 mL, joint: ST/NS 14/23
Expand
View Pricing
Z302759
Quickfit
®
round-bottom flasks,
capacity 250 mL, joint: ST/NS 24/29
Expand
View Pricing
Z302724
Quickfit
®
round-bottom flasks,
capacity 50 mL, joint: ST/NS 24/29
Expand
View Pricing
Z302813
Quickfit
®
round-bottom flasks,
capacity 150 mL, joint: ST/NS 29/32
Expand
View Pricing
Z302856
Quickfit
®
round-bottom flasks,
capacity 1,000 mL, joint: ST/NS 29/32
Expand
View Pricing
Z302651
Quickfit
®
round-bottom flasks,
capacity 100 mL, joint: ST/NS 19/26
Expand
View Pricing
Z302783
Quickfit
®
round-bottom flasks,
capacity 2,000 mL, joint: ST/NS 24/29
Expand
View Pricing
Z302864
Quickfit
®
round-bottom flasks,
capacity 2,000 mL, joint: ST/NS 29/32
Expand
View Pricing
Z302848
Quickfit
®
round-bottom flasks,
capacity 500 mL, joint: ST/NS 29/32
Expand
View Pricing
Z302805
Quickfit
®
round-bottom flasks,
capacity 100 mL, joint: ST/NS 29/32
Expand
View Pricing
Z302678
Quickfit
®
round-bottom flasks,
capacity 150 mL, joint: ST/NS 19/26
Expand
View Pricing
Z302589
Quickfit
®
round-bottom flasks,
capacity 10 mL, joint: ST/NS 14/23
Expand
View Pricing
Z302597
Quickfit
®
round-bottom flasks,
capacity 25 mL, joint: ST/NS 14/23
Expand
View Pricing
Z302627
Quickfit
®
round-bottom flasks,
capacity 250 mL, joint: ST/NS 14/23
Expand
View Pricing
Z302619
Quickfit
®
round-bottom flasks,
capacity 100 mL, joint: ST/NS 14/23
Expand
View Pricing
Page 1 of 2
Page 1 of 2