Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyruvic -2-
13
C acid
Linear Formula:
CH
3
13
COCO
2
H
CAS Number:
70155-58-7
Molecular Weight:
89.05
Products
Product Category
stable isotopes (2)
metabolites (1)
organic acids (1)
Brand
Sigma-Aldrich (1)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Special Grade
GMP (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
692670
Pyruvic -2-
13
C acid,
≥99 atom %
13
C, ≥99% (CP)
Expand
View Pricing
W6624
Pyruvic -2-
13
C acid,
API for Clinical Studies, ≥99 atom %
13
C, ≥95% (CP)
Expand
View Pricing