Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyronin Y
Synonym(s):
Pyronin Y, Pyronin G, Pyronin J
Empirical Formula (Hill Notation):
C
17
H
19
ClN
2
O
CAS Number:
92-32-0
Molecular Weight:
302.80
EC Number:
202-147-8
Products
Product Category
stains (2)
Brand
Sigma-Aldrich (3)
Formula Weight
Melting Point (°C)
Physical Form
Agency
Chemical Composition
Quality Segment
Application
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
P9172
Pyronin Y,
for NA electrophoresis
Expand
View Pricing
83200
Pyronin Y,
Expand
View Pricing
213519
Pyronin Y,
certified by the BSC, Dye content 50 %
Expand
View Pricing