Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
Shorty serological disposable pipette, sterile, plugged
Synonym(s):
Corning
®
7078B
Products
Product Category
pipettes (5)
Manufacturer
Capacity (mL)
Length (mm)
Package Size
Sterility
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
CLS7078B50
Pyrex
®
Shorty serological disposable pipette, sterile, plugged,
volume 50.0 mL, accuracy: 0.5 mL
Expand
View Pricing
CLS7078B5
Pyrex
®
Shorty serological disposable pipette, sterile, plugged,
volume 5 mL, accuracy: 0.1 mL
Expand
View Pricing
CLS7078B25
Pyrex
®
Shorty serological disposable pipette, sterile, plugged,
volume 25.0 mL, accuracy: 0.2 mL
Expand
View Pricing
CLS7078B10
Pyrex
®
Shorty serological disposable pipette, sterile, plugged,
volume 10.0 mL, accuracy: 0.2 mL
Expand
View Pricing
CLS7078B1
Pyrex
®
Shorty serological disposable pipette, sterile, plugged,
volume 1.0 mL, accuracy: 0.01 mL
Expand
View Pricing