
Punicalagin

Empirical Formula (Hill Notation):
C48H28O30
CAS Number:
Molecular Weight:
1084.72
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing