Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
p
-Toluenesulfonic acid monohydrate
Linear Formula:
CH
3
C
6
H
4
SO
3
H · H
2
O
CAS Number:
6192-52-5
Molecular Weight:
190.22
EC Number:
203-180-0
Products
Product Category
organic acids (3)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
ACS reagent (1)
analytical standard (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
27815
p
-Toluenesulfonic acid monohydrate,
≥99% (calc. to H
2
O free subst.)
Expand
View Pricing
402885
p
-Toluenesulfonic acid monohydrate,
ACS reagent, ≥98.5%
Expand
View Pricing
89866
p
-Toluenesulfonic acid monohydrate,
analytical standard
Expand
View Pricing
T35920
p
-Toluenesulfonic acid monohydrate,
ReagentPlus
®
, ≥98%
Expand
View Pricing