Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Probumin
®
Bovine Serum Albumin Crystallized Grade
CAS Number:
9048-46-8
Products
Product Category
albumins (3)
Brand
Sigma-Aldrich (3)
Biological Source
Physical Form
Tissue
Quality Segment
Geographic Origin
Package Size
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
810014
Probumin
®
Bovine Serum Albumin Crystallized Grade,
≥99%, USA Origin, Protein standard: suitable, ligand binding assay: suitable, EIA: Suitable ELISA: suitable
Expand
View Pricing
810016
Probumin
®
Bovine Serum Albumin Crystallized Grade,
≥99%, USA Origin, Protein standard: suitable, ligand binding assay: suitable, EIA: Suitable ELISA: suitable
Expand
View Pricing
810017
Probumin
®
Bovine Serum Albumin Crystallized Grade,
≥99%, USA Origin, Protein standard: suitable, ligand binding assay: suitable, EIA: Suitable ELISA: suitable
Expand
View Pricing