
Prilocaine for peak identification

Synonym(s):
Prilocaine hydrochloride, N-(2-Methylphenyl)-2-(propylamino)propanamide hydrochloride
Empirical Formula (Hill Notation):
C13H20N2O · HCl
CAS Number:
Molecular Weight:
256.77
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing