Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium sulfate
Synonym(s):
Potassium sulfate
Linear Formula:
K
2
SO
4
CAS Number:
7778-80-5
Molecular Weight:
174.26
EC Number:
231-915-5
Products
Product Category
salts (12)
Brand
SAFC (3)
Sigma-Aldrich (8)
Supelco (2)
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Special Grade
ACS reagent (2)
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
P0772
Potassium sulfate,
ReagentPlus
®
, ≥99.0%
Expand
View Pricing
1.05153
Potassium sulfate,
for analysis EMSURE
®
ACS,ISO,Reag. Ph Eur
Expand
View Pricing
223492
Potassium sulfate,
ACS reagent, ≥99.0%, powder
Expand
View Pricing
221325
Potassium sulfate,
ACS reagent, ≥99.0%, powder or crystals
Expand
View Pricing
1.05151
Potassium sulfate,
powder, EMPROVE
®
ESSENTIAL, Ph. Eur., BP, JP, E 515
Expand
View Pricing
12658
Potassium sulfate,
meets analytical specification of Ph. Eur., 99-101%
Expand
View Pricing
204129
Potassium sulfate,
99.99% trace metals basis
Expand
View Pricing
1.05152
Potassium sulfate,
99.999 Suprapur
®
Expand
View Pricing
60528
Potassium sulfate,
BioUltra, ≥99.0% (AT)
Expand
View Pricing
1.05150
Potassium sulfate,
crystalline, EMPROVE
®
ESSENTIAL, Ph. Eur., BP, JP
Expand
View Pricing
P8541
Potassium sulfate,
suitable for plant cell culture
Expand
View Pricing
P9458
Potassium sulfate,
BioXtra, ≥99.0%
Expand
View Pricing
RES0797P-A7
Potassium sulfate,
Expand
View Pricing