Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Portland Cement
Synonym(s):
Portland cement
EC Number:
266-043-4
Products
Manufacturer
Physical Form
Format
Quality Segment
Reference Material Type
Special Grade
certified reference material (10)
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
NIST114R
Portland Cement,
NIST
®
SRM
®
114r, fineness standard
Expand
View Pricing
NIST46H
Portland Cement,
NIST
®
SRM
®
46h, fineness standard
Expand
View Pricing
NIST1881B
Portland Cement,
Blended with Fly Ash, NIST
®
SRM
®
1881B
Expand
View Pricing
NIST1880B
Portland Cement,
NIST
®
SRM
®
1880b
Expand
View Pricing
NIST1884B
Portland Cement,
NIST
®
SRM
®
1884b
Expand
View Pricing
NIST633A
Portland Cement,
NIST
®
SRM
®
633a
Expand
View Pricing
NIST1885B
Portland Cement,
NIST
®
SRM
®
1885a
Expand
View Pricing
NIST1888B
Portland Cement,
NIST
®
SRM
®
1888b
Expand
View Pricing
NIST1887B
Portland Cement,
NIST
®
SRM
®
1887b
Expand
View Pricing
NIST634A
Portland Cement,
NIST
®
SRM
®
634a
Expand
View Pricing
NIST1886A
Portland Cement,
NIST
®
SRM
®
1886a, white portland cement with low iron
Expand
View Pricing