
Poly(tetrahydrofuran)

Synonym(s):
α-Hydro-ω-hydroxypoly(oxy-1,4-butanediyl), Poly(1,4-butanediol), polyTHF
Linear Formula:
H(OCH2CH2CH2CH2)nOH
CAS Number:
Products
Brand
Melting Point (°C)
Physical Form
Quality Segment
Polymer Composition
Polymer Type
Showing 1-3 of 3