Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(styrenesulfonic acid sodium salt)
Synonym(s):
Poly(sodium styrenesulfonate)
CAS Number:
9080-79-9
Products
Brand
Supelco (7)
Format
Quality Segment
Special Grade
analytical standard (7)
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
81609
Poly(styrenesulfonic acid sodium salt),
analytical standard, for GPC, 17,000
Expand
View Pricing
81606
Poly(styrenesulfonic acid sodium salt),
analytical standard, for GPC, 4,300
Expand
View Pricing
81614
Poly(styrenesulfonic acid sodium salt),
analytical standard, for GPC, 150,000
Expand
View Pricing
81604
Poly(styrenesulfonic acid sodium salt),
analytical standard, for GPC, 210
Expand
View Pricing
81612
Poly(styrenesulfonic acid sodium salt),
analytical standard, for GPC, 77,000
Expand
View Pricing
81610
Poly(styrenesulfonic acid sodium salt),
analytical standard, for GPC, 32,000
Expand
View Pricing
81607
Poly(styrenesulfonic acid sodium salt),
analytical standard, for GPC, 6,800
Expand
View Pricing