Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(ethylene oxide)
Synonym(s):
PEO, Poly(ethylene glycol), PEG
Linear Formula:
H(OCH
2
CH
2
)
n
OH
CAS Number:
25322-68-3
Products
Brand
Sigma-Aldrich (11)
Physical Form
Quality Segment
Functional Group
Application
Polymer Composition
Polymer Type
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
181986
Poly(ethylene oxide),
average M
V
100,000 (nominal), powder, hydroxyl, BHT as inhibitor
Expand
View Pricing
182028
Poly(ethylene oxide),
average M
V
600,000 (nominal), powder, hydroxyl, BHT as inhibitor
Expand
View Pricing
189464
Poly(ethylene oxide),
average M
V
4,000,000 (nominal), powder, hydroxyl, BHT as inhibitor
Expand
View Pricing
372781
Poly(ethylene oxide),
average M
V
1,000,000 (nominal), powder, hydroxyl, BHT as inhibitor
Expand
View Pricing
181994
Poly(ethylene oxide),
average M
V
200,000 (nominal), powder, hydroxyl, BHT as inhibitor
Expand
View Pricing
372773
Poly(ethylene oxide),
average M
V
400,000 (nominal), powder, hydroxyl, BHT as inhibitor
Expand
View Pricing
182001
Poly(ethylene oxide),
average M
V
300,000 (nominal), powder, hydroxyl, BHT as inhibitor
Expand
View Pricing
189456
Poly(ethylene oxide),
average M
V
900,000 (nominal), powder, hydroxyl, BHT as inhibitor
Expand
View Pricing
189472
Poly(ethylene oxide),
average M
V
5,000,000 (nominal), powder, hydroxyl, BHT as inhibitor
Expand
View Pricing
372803
Poly(ethylene oxide),
average M
V
2,000,000 (nominal), powder, hydroxyl, BHT as inhibitor
Expand
View Pricing
372838
Poly(ethylene oxide),
average M
V
8,000,000 (nominal), powder, hydroxyl, BHT as inhibitor
Expand
View Pricing
1546853
Poly(ethylene oxide),
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing