Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Polyethylene glycol solution
Synonym(s):
PEG solution
CAS Number:
25322-68-3
Products
Product Category
detergents (2)
Brand
Sigma-Aldrich (1)
pH Value
Physical Form
Product Line
Quality Segment
Sterilization Method
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
76293
Polyethylene glycol solution,
BioUltra, for molecular biology, 1,000, ~50% in H
2
O
Expand
View Pricing
P1458
Polyethylene glycol solution,
40 % (w/w) in H
2
O, average mol wt 8,000
Expand
View Pricing