Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Polyethylene
Linear Formula:
H(CH
2
CH
2
)
n
H
CAS Number:
9002-88-4
Products
Brand
Sigma-Aldrich (8)
Supelco (1)
Melting Point (°C)
Physical Form
Quality Segment
Particle Size (µm)
Polymer Composition
Polymer Type
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
434272
Polyethylene,
Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Expand
View Pricing
427772
Polyethylene,
average M
w
~4,000 by GPC, average M
n
~1,700 by GPC
Expand
View Pricing
428043
Polyethylene,
low density, melt index 25 g/10 min (190°C/2.16kg)
Expand
View Pricing
429015
Polyethylene,
Ultra-high molecular weight, average M
w
3,000,000-6,000,000
Expand
View Pricing
332119
Polyethylene,
Medium density
Expand
View Pricing
427985
Polyethylene,
High density, melt index 12 g/10 min (190 °C/2.16kg)
Expand
View Pricing
428078
Polyethylene,
Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Expand
View Pricing
434264
Polyethylene,
Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Expand
View Pricing
547999
Polyethylene,
High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Expand
View Pricing
81223
Polyethylene,
analytical standard, for GPC, 2,000
Expand
View Pricing