Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(ε-caprolactone)-
b
-poly(ethylene glycol)-
b
-poly(ε-caprolactone)
Linear Formula:
H(OCH
2
CH
2
CH
2
CH
2
CH
2
CO)
m
O(CH
2
CH
2
O)
n
(COCH
2
CH
2
CH
2
CH
2
CH
2
O)
m
H
Products
Brand
Sigma-Aldrich (3)
Physical Form
Quality Segment
Polymer Composition
Polymer Type
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
900669
Poly(ε-caprolactone)-
b
-poly(ethylene glycol)-
b
-poly(ε-caprolactone),
6k-6k-6k
Expand
View Pricing
900653
Poly(ε-caprolactone)-
b
-poly(ethylene glycol)-
b
-poly(ε-caprolactone),
5k-10k-5k
Expand
View Pricing
900670
Poly(ε-caprolactone)-
b
-poly(ethylene glycol)-
b
-poly(ε-caprolactone),
2k-2k-2k
Expand
View Pricing