Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Platinum/Rhodium
Products
Manufacturer
Physical Form
Length (m)
Filter Products
Showing 1-20 of 33
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF13464457
Platinum/Rhodium,
wire, Pt 80%/Rh 20%, 0.25 mm diameter, length 0.05 m
Expand
View Pricing
GF43012145
Platinum/Rhodium,
wire, Pt 80%/Rh 20%, 0.5 mm diameter, length 0.2 m
Expand
View Pricing
GF17267991
Platinum/Rhodium,
wire, Pt 90%/Rh10%, 0.38 mm diameter, length 0.05 m, temper annealed
Expand
View Pricing
GF70529104
Platinum/Rhodium,
wire, 0.015 mm diameter, Pt87%/Rh13%, length 5 m, temper annealed
Expand
View Pricing
GF00403962
Platinum/Rhodium,
tube, Pt 90%/Rh10%, outside diameter 3.18 mm, length 50 mm, wall thickness 0.125 mm
Expand
View Pricing
GF20654232
Platinum/Rhodium,
wire, Pt 90%/Rh10%, 0.38 mm diameter, length 0.5 m, temper annealed
Expand
View Pricing
GF47507834
Platinum/Rhodium,
wire, Pt 80%/Rh 20%, 0.25 mm diameter, length 0.2 m
Expand
View Pricing
GF11207988
Platinum/Rhodium,
wire, Pt 70%/Rh 30%, 0.025 mm diameter, length 2 m, temper as drawn
Expand
View Pricing
GF54627611
Platinum/Rhodium,
tube, Pt 90%/Rh10%, outside diameter 3.18 mm, length 100 mm, wall thickness 0.125 mm
Expand
View Pricing
GF09101865
Platinum/Rhodium,
tube, Pt 90%/Rh10%, outside diameter 1.6 mm, length 100 mm, wall thickness 0.3 mm
Expand
View Pricing
GF30294446
Platinum/Rhodium,
wire, Pt 80%/Rh 20%, 0.5 mm diameter, length 0.1 m
Expand
View Pricing
GF37251277
Platinum/Rhodium,
wire, Pt 90%/Rh10%, 0.38 mm diameter, length 0.1 m, temper annealed
Expand
View Pricing
GF06984142
Platinum/Rhodium,
tube, Pt 90%/Rh10%, outside diameter 1.6 mm, length 50 mm, wall thickness 0.3 mm
Expand
View Pricing
GF72700196
Platinum/Rhodium,
wire, Pt94%/Rh 6%, 0.50 mm diameter, length 0.05 m, temper annealed
Expand
View Pricing
GF96383807
Platinum/Rhodium,
wire, 0.25 mm diameter, Pt80%/Rh20%, length 1 m
Expand
View Pricing
GF16682902
Platinum/Rhodium,
wire, Pt 94%/Rh 6%, 1.0 mm diameter, length 0.1 m, temper annealed
Expand
View Pricing
GF74212152
Platinum/Rhodium,
foil, thickness 0.125 mm, Pt60%/Rh40%, size 25 × 25 mm, temper as rolled
Expand
View Pricing
GF69897571
Platinum/Rhodium,
wire, 0.38 mm diameter, Pt90%/Rh10%, length 0.2 m, temper annealed
Expand
View Pricing
GF83917109
Platinum/Rhodium,
wire, Pt70%/Rh30%, 0.025 mm diameter, length 0.5 m, temper as drawn
Expand
View Pricing
GF02644400
Platinum/Rhodium,
wire, Pt 70%/Rh 30%, 0.025 mm diameter, length 5 m, temper as drawn
Expand
View Pricing
Page 1 of 2
Page 1 of 2