Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Platinum-iridium alloy, Pt90Ir10
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-20 of 82
Page 1 of 5
Page 1 of 5
Compare
Product Number
Description
Pricing
GF18233052
Platinum-iridium alloy, Pt90Ir10,
foil, 50 x 50mm, 0.125mm thickness, as rolled
Expand
View Pricing
GF93124908
Platinum-iridium alloy, Pt90Ir10,
foil, 25 x 25mm, 0.125mm thickness, as rolled
Expand
View Pricing
GF67002757
Platinum-iridium alloy, Pt90Ir10,
200mm tube, 0.56mm outside diameter, 0.13mm wall thickness, 0.30mm inside diameter, as drawn
Expand
View Pricing
GF89509887
Platinum-iridium alloy, Pt90Ir10,
50mm rod, 2.0mm diameter, as drawn
Expand
View Pricing
GF59941143
Platinum-iridium alloy, Pt90Ir10,
1m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF55986261
Platinum-iridium alloy, Pt90Ir10,
50mm tube, 0.56mm outside diameter, 0.13mm wall thickness, 0.30mm inside diameter, as drawn
Expand
View Pricing
GF39013040
Platinum-iridium alloy, Pt90Ir10,
0.5m wire, 0.5mm diameter, as drawn
Expand
View Pricing
GF19916073
Platinum-iridium alloy, Pt90Ir10,
25mm rod, 2.0mm diameter, as drawn
Expand
View Pricing
GF88735225
Platinum-iridium alloy, Pt90Ir10,
100mm rod, 2.0mm diameter, as drawn
Expand
View Pricing
GF11736020
Platinum-iridium alloy, Pt90Ir10,
100mm tube, 0.56mm outside diameter, 0.13mm wall thickness, 0.30mm inside diameter, as drawn
Expand
View Pricing
GF98479827
Platinum-iridium alloy, Pt90Ir10,
foil, 25 x 25mm, 0.10mm thickness, as rolled
Expand
View Pricing
GF59759937
Platinum-iridium alloy, Pt90Ir10,
0.2m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF93322010
Platinum-iridium alloy, Pt90Ir10,
0.5m wire, 0.125mm diameter, as drawn
Expand
View Pricing
GF34911121
Platinum-iridium alloy, Pt90Ir10,
0.1m wire, 0.1mm diameter, as drawn
Expand
View Pricing
GF99895033
Platinum-iridium alloy, Pt90Ir10,
0.1m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF27946570
Platinum-iridium alloy, Pt90Ir10,
0.1m wire, 0.125mm diameter, as drawn
Expand
View Pricing
GF65337666
Platinum-iridium alloy, Pt90Ir10,
foil, 25 x 25mm, 0.025mm thickness, as rolled, not light tested (nlt)
Expand
View Pricing
GF63432930
Platinum-iridium alloy, Pt90Ir10,
foil, 25 x 25mm, 0.25mm thickness, as rolled
Expand
View Pricing
GF44346359
Platinum-iridium alloy, Pt90Ir10,
foil, 50 x 50mm, 0.05mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF35380874
Platinum-iridium alloy, Pt90Ir10,
1m wire, 0.38mm diameter, as drawn
Expand
View Pricing
Page 1 of 5
Page 1 of 5