Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Platinum black
Empirical Formula (Hill Notation):
Pt
CAS Number:
7440-06-4
Molecular Weight:
195.08
EC Number:
231-116-1
Products
Brand
Sigma-Aldrich (3)
Sustainability
Feature
Formula Weight
Physical Form
Purity
Particle Size (µm)
Reagent Type
Core
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
205915
Platinum black,
black, powder, ≤20 μm, ≥99.95% trace metals basis
Expand
View Pricing
520780
Platinum black,
fuel cell grade, ≥99.9% trace metals basis
Expand
View Pricing
520799
Platinum black,
low bulk density, ≥99.9% trace metals basis
Expand
View Pricing