Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Phosphor bronze
Products
Manufacturer
Physical Form
Format
Length (m)
Quality Segment
Reference Material Type
Special Grade
certified reference material (2)
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
GF14135138
Phosphor bronze,
powder, Cu 94%/Sn 6%, max. particle size 150 micron, weight 100 g, condition alloy pre-cursor
Expand
View Pricing
GF66891561
Phosphor bronze,
powder, Cu94%/Sn 6%, 150 max. part. size (micron), condition alloy pre-cursor, weight 200 g
Expand
View Pricing
GF91109257
Phosphor bronze,
foil, thickness 0.08 mm, Cu94%/Sn 6%, length 10 m, coil width 0.4 mm
Expand
View Pricing
GF55511524
Phosphor bronze,
foil, Cu 94%/Sn 6%, thickness 0.08 mm, length 0.5 m, coil width 0.4 mm
Expand
View Pricing
GF98025891
Phosphor bronze,
powder, Cu94%/Sn 6%, 150 max. part. size (micron), weight 50 g, condition alloy pre-cursor
Expand
View Pricing
GF59643681
Phosphor bronze,
foil, thickness 0.08 mm, Cu94%/Sn 6%, length 2 m, coil width 0.4 mm
Expand
View Pricing
GF77502304
Phosphor bronze,
foil, thickness 0.08 mm, Cu94%/Sn 6%, length 1 m, coil width 0.4 mm
Expand
View Pricing
NIST872
Phosphor bronze,
NIST
®
SRM
®
872,(CDA 544)
Expand
View Pricing
GF77146735
Phosphor bronze,
foil, Cu94%/Sn 6%, thickness 0.08 mm, length 5 m, coil width 0.4 mm
Expand
View Pricing
NIST871
Phosphor bronze,
NIST
®
SRM
®
871, (CDA 521)
Expand
View Pricing