Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Peppermint oil
Synonym(s):
Mentha piperita
CAS Number:
8006-90-4
Products
Product Category
essential oils (2)
Brand
Sigma-Aldrich (4)
Biological Source
Boiling Point (°C)
Physical Form
Agency
Quality Segment
Geographic Origin
Application
Organoleptic Class
Special Grade
Kosher (3)
Food Grade (2)
Halal (1)
Natural (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
77411
Peppermint oil,
natural, from
Mentha piperita
L.
Expand
View Pricing
W284815
Peppermint oil,
natural, FCC, FG
Expand
View Pricing
W284823
Peppermint oil,
purified by triple-distillation, FCC, FG
Expand
View Pricing
W284831
Peppermint oil,
terpeneless, FCC
Expand
View Pricing