Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Palladium on carbon
Linear Formula:
Pd
Products
Brand
Sigma-Aldrich (8)
Physical Form
Element
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
908029
Palladium on carbon,
Evonik Noblyst
®
P1141 10% Pd
Expand
View Pricing
907987
Palladium on carbon,
Evonik Noblyst
®
P1095 5% Pd
Expand
View Pricing
907839
Palladium on carbon,
Evonik Noblyst
®
P1090 5% Pd
Expand
View Pricing
908118
Palladium on carbon,
Evonik Noblyst
®
P1145 5% Pd
Expand
View Pricing
907944
Palladium on carbon,
Evonik Noblyst
®
P1092 5% Pd
Expand
View Pricing
908096
Palladium on carbon,
Evonik Noblyst
®
P1126 5% Pd
Expand
View Pricing
907995
Palladium on carbon,
Evonik Noblyst
®
P1109 5% Pd
Expand
View Pricing
908061
Palladium on carbon,
Evonik Noblyst
®
P1093 5% Pd
Expand
View Pricing