Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Oxidizer
Synonym(s):
Oxidizer
Products
Product Category
nucleosides (4)
Brand
Sigma-Aldrich (7)
Color
Physical Form
Product Line
Quality Segment
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
L060050
Oxidizer,
0.05 M, configured for (ÄKTA
®
and OligoPilot
®
)
Expand
View Pricing
L060030
Oxidizer,
0.1 M, configured for ABI
Expand
View Pricing
L060080
Oxidizer,
0.02 M, configured for PerkinElmer, configured for Polygen
Expand
View Pricing
L060020
Oxidizer,
0.02 M, configured for ABI 3900
Expand
View Pricing
L060060
Oxidizer,
0.02 M, configured for ABI
Expand
View Pricing
L060084
Oxidizer,
Expand
View Pricing
L060005
Oxidizer,
Expand
View Pricing