
Oleandrin

Synonym(s):
5β,20(22)-Cardenolide-3β,14,16β-triol-3-([2,6-dideoxy-3-O-methyl-α-L-arabinohexopyranosyl]oxy) 16-acetate, Corrigen, Foliandrin, Folinerin, Neriol, Neriolin, Neriostene
Empirical Formula (Hill Notation):
C32H48O9
CAS Number:
Molecular Weight:
576.72
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing