
Olanzapine for system suitability

Synonym(s):
Olanzapine, 2-Methyl-4-(4-methyl-1-piperazinyl)- 10H-thieno[2,3-b][1,5]benzodiazepine
Empirical Formula (Hill Notation):
C17H20N4S
CAS Number:
Molecular Weight:
312.43
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing