Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ohaus
®
Pioneer
®
Precision Balance
Products
Product Category
balances (9)
Manufacturer
Weighing Capacity (g)
Resolution (g)
Package Size
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
OH30429812
Ohaus
®
Pioneer
®
Precision Balance,
Model PX323/E, weighing capacity 320 g, resolution: 0.001 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30429814
Ohaus
®
Pioneer
®
Precision Balance,
Model PX3202/E, weighing capacity 3200 g, resolution: 0.01 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30429816
Ohaus
®
Pioneer
®
Precision Balance,
Model PX4201/E, weighing capacity 4200 g, resolution: 0.1 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30429805
Ohaus
®
Pioneer
®
Precision Balance,
Model PX323, weighing capacity 320 g, resolution: 0.001 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30430015
Ohaus
®
Pioneer
®
Precision Balance,
Model PX323M, weighing capacity 320 g, resolution: 0.001 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30429807
Ohaus
®
Pioneer
®
Precision Balance,
Model PX3202, weighing capacity 3200 g, resolution: 0.01 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30429809
Ohaus
®
Pioneer
®
Precision Balance,
Model PX4201, weighing capacity 4200 g, resolution: 0.1 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30430021
Ohaus
®
Pioneer
®
Precision Balance,
Model PX3202M, weighing capacity 3200 g, resolution: 0.01 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30430024
Ohaus
®
Pioneer
®
Precision Balance,
Model PX4201M, weighing capacity 4200 g, resolution: 0.1 g, AC/DC input 230 V, universal plug set
Expand
View Pricing