Skip to Content
Merck
Octanoic acid analytical standard

Octanoic acid

Octanoic acid analytical standard
Synonym(s):
Acid C8, Caprylic acid, Octanoic acid
Linear Formula:
CH3(CH2)6COOH
CAS Number:
Molecular Weight:
144.21
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing