Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nunc
®
MicroWell
™
96 well polystyrene plates
Products
Product Category
plates (9)
Color
Manufacturer
Binding Type
Length (mm)
Volume (µL)
Wells
Color
Package Size
Sterility
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
P7366
Nunc
®
MicroWell
™
96 well polystyrene plates,
polystyrene, clear, flat bottom, 400 μL/well, non sterile, 60/cs
Expand
View Pricing
P8741
Nunc
®
MicroWell
™
96 well polystyrene plates,
Nunclon
®
Delta, black, well volume 400 μL, case of 50 ea
Expand
View Pricing
P8616
Nunc
®
MicroWell
™
96 well polystyrene plates,
Nunclon
®
Delta, white, well volume 400 μL, case of 50 ea
Expand
View Pricing
P5366
Nunc
®
MicroWell
™
96 well polystyrene plates,
polystyrene, clear, round bottom, 300 μL/well, sterile, 50/cs
Expand
View Pricing
P7491
Nunc
®
MicroWell
™
96 well polystyrene plates,
polystyrene, clear, flat bottom, 400 μL/well, sterile, 50/cs
Expand
View Pricing
Z688665
Nunc
®
MicroWell
™
96 well polystyrene plates,
size 96 well, conical clear wells, non-treated surface, non-sterile, case of 180
Expand
View Pricing
P4241
Nunc
®
MicroWell
™
96 well polystyrene plates,
polystyrene, clear, conical bottom, 300 μL/well, sterile, 50/cs
Expand
View Pricing
TMO137103
Nunc
®
MicroWell
™
96 well polystyrene plates,
Nunclon
®
Delta, black, well volume (400 μL), case of 160 ea
Expand
View Pricing
TMO136102
Nunc
®
MicroWell
™
96 well polystyrene plates,
Nunclon
®
Delta, white, well volume (400 μL), case of 160 ea
Expand
View Pricing