Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nunc
®
Coded Cryobank Vials in black low rack
Products
Product Category
vials (8)
Manufacturer
Capacity (mL)
Color
Package Size
Special Grade
Natural (8)
Sterility
Technique
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
TMO374088
Nunc
®
Coded Cryobank Vials in black low rack,
polypropylene, capacity 1.0 mL, self-standing natural, sterile, clear cap, case of 960 ea
Expand
View Pricing
TMO374086
Nunc
®
Coded Cryobank Vials in black low rack,
polypropylene, capacity 0.5 mL, self-standing natural, sterile, clear cap, case of 960 ea
Expand
View Pricing
TMO374027
Nunc
®
Coded Cryobank Vials in black low rack,
polypropylene, capacity 0.5 mL, self-standing natural, sterile, green cap, case of 960 ea
Expand
View Pricing
TMO374025
Nunc
®
Coded Cryobank Vials in black low rack,
polypropylene, capacity 0.5 mL, self-standing natural, sterile, blue cap, case of 960 ea
Expand
View Pricing
TMO374110
Nunc
®
Coded Cryobank Vials in black low rack,
polypropylene, capacity 1.0 mL, self-standing natural, sterile, green cap, case of 960 ea
Expand
View Pricing
TMO374026
Nunc
®
Coded Cryobank Vials in black low rack,
polypropylene, capacity 0.5 mL, self-standing natural, sterile, red cap, case of 960 ea
Expand
View Pricing
TMO374130
Nunc
®
Coded Cryobank Vials in black low rack,
polypropylene, capacity 1.0 mL, self-standing natural, sterile, blue cap, case of 960 ea
Expand
View Pricing
TMO374120
Nunc
®
Coded Cryobank Vials in black low rack,
polypropylene, capacity 1.0 mL, self-standing natural, sterile, red cap, case of 960 ea
Expand
View Pricing