Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nunc
®
Cell Factory, Nunclon
®
Δ surface
Products
Color
Manufacturer
Surface Area cm²
Working Volume (ml)
Package Size
Sterility
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
Z720895
Nunc
®
Cell Factory, Nunclon
®
Δ surface,
10-chamber CF, 6320 cm
2
, 2000 mL, 6/cs
Expand
View Pricing
Z720860
Nunc
®
Cell Factory, Nunclon
®
Δ surface,
4-chamber CF, 2528 cm
2
, 800 mL, 10/cs
Expand
View Pricing
Z720887
Nunc
®
Cell Factory, Nunclon
®
Δ surface,
2-chamber CF, 1264 cm
2
, 400 mL, 6/cs
Expand
View Pricing
TMO164327
Nunc
®
Cell Factory, Nunclon
®
Δ surface,
Nunclon
®
Delta surface, 10 layers, polystyrene, surface area (6320 cm
2
), case of 2 ea
Expand
View Pricing
Z720879
Nunc
®
Cell Factory, Nunclon
®
Δ surface,
1-chamber CF, 632 cm
2
, 200 mL, 8/cs
Expand
View Pricing