Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
N
-
tert
-Butyl-α-phenylnitrone
Synonym(s):
N
-Benzylidene-
tert
-butylamine N-oxide, PBN, Phenyl
N
-t-butylnitrone, N-
tert
-Butyl-α-phenylnitrone, N-tert-Butyl-α-phenylnitrone
Linear Formula:
C
6
H
5
CH=N(O)C(CH
3
)
3
CAS Number:
3376-24-7
Molecular Weight:
177.24
EC Number:
222-168-6
Products
Product Category
bioactive small molecules (1)
inhibitors (1)
Brand
Sigma-Aldrich (3)
Supelco (1)
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Research Area
Quality Segment
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
B7263
N
-
tert
-Butyl-α-phenylnitrone,
≥98% (GC)
Expand
View Pricing
203995
N
-
tert
-Butyl-α-phenylnitrone,
Most commonly-used free radical trap.
Expand
View Pricing
8.41179
N
-
tert
-Butyl-α-phenylnitrone,
for synthesis
Expand
View Pricing
80126
N
-
tert
-Butyl-α-phenylnitrone,
for ESR-spectroscopy
Expand
View Pricing