Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Norell
®
valved 5 mm NMR tubes for intermediate pressure
Products
Product Category
tubes (12)
Manufacturer
Diameter (mm)
Length (in)
Frequency (MHz)
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
NRS5500MWIPV7
Norell
®
valved 5 mm NMR tubes for intermediate pressure,
frequency 500 MHz, L 7 in., medium wall
Expand
View Pricing
NRS5500HWIPV7
Norell
®
valved 5 mm NMR tubes for intermediate pressure,
frequency 500 MHz, L 7 in., heavy wall
Expand
View Pricing
NRS5500MWIPV8
Norell
®
valved 5 mm NMR tubes for intermediate pressure,
frequency 500 MHz, L 8 in., medium wall
Expand
View Pricing
NRS5500HWIPV8
Norell
®
valved 5 mm NMR tubes for intermediate pressure,
frequency 500 MHz, L 8 in., heavy wall
Expand
View Pricing
NRS5500IPV8
Norell
®
valved 5 mm NMR tubes for intermediate pressure,
frequency 500 MHz, L 8 in., thin wall
Expand
View Pricing
NRS5600IPV7
Norell
®
valved 5 mm NMR tubes for intermediate pressure,
frequency 600 MHz, L 7 in., thin wall
Expand
View Pricing
NRS5600MWIPV7
Norell
®
valved 5 mm NMR tubes for intermediate pressure,
frequency 600 MHz, L 7 in., medium wall
Expand
View Pricing
NRS5600IPV8
Norell
®
valved 5 mm NMR tubes for intermediate pressure,
frequency 600 MHz, L 8 in., thin wall
Expand
View Pricing
NRS5500IPV7
Norell
®
valved 5 mm NMR tubes for intermediate pressure,
frequency 500 MHz, L 7 in., thin wall
Expand
View Pricing
NRS5600HWIPV7
Norell
®
valved 5 mm NMR tubes for intermediate pressure,
frequency 600 MHz, L 7 in., heavy wall
Expand
View Pricing
NRS5600MWIPV8
Norell
®
valved 5 mm NMR tubes for intermediate pressure,
frequency 600 MHz, L 8 in., medium wall
Expand
View Pricing
NRS5600HWIPV8
Norell
®
valved 5 mm NMR tubes for intermediate pressure,
frequency 600 MHz, L 8 in., heavy wall
Expand
View Pricing