Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
N,N
-Dimethyltrimethylsilylamine
Synonym(s):
(Dimethylamino)trimethylsilane,
N
-(Trimethylsilyl)dimethylamine, TMSDMA
Linear Formula:
(CH
3
)
3
SiN(CH
3
)
2
CAS Number:
2083-91-2
Molecular Weight:
117.26
EC Number:
218-222-3
Products
Product Category
building blocks (1)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
226289
N,N
-Dimethyltrimethylsilylamine,
97%
Expand
View Pricing