Skip to Content
Merck
N,N′-Di(1-naphthyl)-N,N′-diphenyl-(1,1′-biphenyl)-4,4′-diamine 96%

N,N′-Di(1-naphthyl)-N,N′-diphenyl-(1,1′-biphenyl)-4,4′-diamine

N,N′-Di(1-naphthyl)-N,N′-diphenyl-(1,1′-biphenyl)-4,4′-diamine 96%
Synonym(s):
N,N′-Bis(naphthalen-1-yl)-N,N′-bis(phenyl)benzidine, NPB, NPD
Empirical Formula (Hill Notation):
C44H32N2
CAS Number:
Molecular Weight:
588.74

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Special Grade