Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
N
-Methyl-bis(trifluoroacetamide)
Synonym(s):
MBTFA
Linear Formula:
CF
3
CON(CH
3
)COCF
3
CAS Number:
685-27-8
Molecular Weight:
223.07
EC Number:
211-680-5
Products
Brand
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Special Grade
Puriss (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
M0789
N
-Methyl-bis(trifluoroacetamide),
derivatization grade (GC derivatization), LiChropur
™
, ≥97.0% (GC)
Expand
View Pricing
65943
N
-Methyl-bis(trifluoroacetamide),
derivatization grade (GC derivatization), LiChropur
™
, ≥99.0% (GC)
Expand
View Pricing