Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nile Blue A
Synonym(s):
Basic Blue 12, Nile blue sulfate
Linear Formula:
2C
20
H
20
N
3
O · SO
4
CAS Number:
3625-57-8
Molecular Weight:
732.85
EC Number:
222-832-5
Products
Product Category
stains (2)
Brand
Sigma-Aldrich (2)
Color
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Application
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
N5632
Nile Blue A,
Dye content ≥75 %, Powder
Expand
View Pricing
N0766
Nile Blue A,
Dye content, ≥75%, certified by the Biological Stain Commission, powder
Expand
View Pricing