
(−)-Nicotine hemisulfate salt

Synonym(s):
(−)-1-Methyl-2-(3-pyridyl)pyrrolidine
Linear Formula:
(C10H14N2)2 · H2SO4
CAS Number:
Molecular Weight:
422.54
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing