Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nickel silver (18) alloy, Cu65Ni18Zn17
Products
Manufacturer
Physical Form
ID (mm)
Length (mm)
OD (mm)
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
GF32725133
Nickel silver (18) alloy, Cu65Ni18Zn17,
100mm tube, 1.6mm outside diameter, 0.15mm wall thickness, 1.3mm inside diameter, hard
Expand
View Pricing
GF92840867
Nickel silver (18) alloy, Cu65Ni18Zn17,
200mm tube, 6.35mm outside diameter, 0.5mm wall thickness, 5.35mm inside diameter, hard
Expand
View Pricing
GF07415917
Nickel silver (18) alloy, Cu65Ni18Zn17,
1000mm tube, 4.0mm outside diameter, 0.25mm wall thickness, 3.5mm inside diameter, hard
Expand
View Pricing
GF35975605
Nickel silver (18) alloy, Cu65Ni18Zn17,
500mm tube, 1.0mm outside diameter, 0.1mm wall thickness, 0.8mm inside diameter, hard
Expand
View Pricing
GF24471693
Nickel silver (18) alloy, Cu65Ni18Zn17,
200mm tube, 1.0mm outside diameter, 0.1mm wall thickness, 0.8mm inside diameter, hard
Expand
View Pricing
GF95760782
Nickel silver (18) alloy, Cu65Ni18Zn17,
100mm tube, 4.0mm outside diameter, 0.25mm wall thickness, 3.5mm inside diameter, hard
Expand
View Pricing
GF73274722
Nickel silver (18) alloy, Cu65Ni18Zn17,
200mm tube, 1.6mm outside diameter, 0.15mm wall thickness, 1.3mm inside diameter, hard
Expand
View Pricing
GF08566207
Nickel silver (18) alloy, Cu65Ni18Zn17,
200mm tube, 4.0mm outside diameter, 0.25mm wall thickness, 3.5mm inside diameter, hard
Expand
View Pricing
GF15333375
Nickel silver (18) alloy, Cu65Ni18Zn17,
100mm tube, 6.35mm outside diameter, 0.5mm wall thickness, 5.35mm inside diameter, hard
Expand
View Pricing
GF68606837
Nickel silver (18) alloy, Cu65Ni18Zn17,
1000mm tube, 1.6mm outside diameter, 0.15mm wall thickness, 1.3mm inside diameter, hard
Expand
View Pricing
GF16722761
Nickel silver (18) alloy, Cu65Ni18Zn17,
500mm tube, 1.6mm outside diameter, 0.15mm wall thickness, 1.3mm inside diameter, hard
Expand
View Pricing
GF39918482
Nickel silver (18) alloy, Cu65Ni18Zn17,
100mm tube, 1.0mm outside diameter, 0.1mm wall thickness, 0.8mm inside diameter, hard
Expand
View Pricing
GF27696213
Nickel silver (18) alloy, Cu65Ni18Zn17,
500mm tube, 6.35mm outside diameter, 0.5mm wall thickness, 5.35mm inside diameter, hard
Expand
View Pricing
GF40537065
Nickel silver (18) alloy, Cu65Ni18Zn17,
1000mm tube, 1.0mm outside diameter, 0.1mm wall thickness, 0.8mm inside diameter, hard
Expand
View Pricing
GF78040728
Nickel silver (18) alloy, Cu65Ni18Zn17,
1000mm tube, 6.35mm outside diameter, 0.5mm wall thickness, 5.35mm inside diameter, hard
Expand
View Pricing