Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nickel-iron alloy, Ni80Fe20
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Filter Products
Showing 1-20 of 28
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF66076229
Nickel-iron alloy, Ni80Fe20,
foil, 100 x 100mm, 1.0mm thickness, as rolled
Expand
View Pricing
GF19623955
Nickel-iron alloy, Ni80Fe20,
foil, 25 x 25mm, 1.0mm thickness, as rolled
Expand
View Pricing
GF58487170
Nickel-iron alloy, Ni80Fe20,
foil, 25 x 25mm, 3.0mm thickness, as rolled
Expand
View Pricing
GF27848054
Nickel-iron alloy, Ni80Fe20,
50g lump, 5mm max. lump size, pellets
Expand
View Pricing
GF68252148
Nickel-iron alloy, Ni80Fe20,
foil, 50 x 50mm, 1.0mm thickness, as rolled
Expand
View Pricing
GF73231376
Nickel-iron alloy, Ni80Fe20,
100mm rod, 2.0mm diameter, as drawn
Expand
View Pricing
GF87062328
Nickel-iron alloy, Ni80Fe20,
100g lump, 5mm max. lump size, pellets
Expand
View Pricing
GF62636519
Nickel-iron alloy, Ni80Fe20,
1m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF53922304
Nickel-iron alloy, Ni80Fe20,
200mm rod, 2.0mm diameter, as drawn
Expand
View Pricing
GF95484550
Nickel-iron alloy, Ni80Fe20,
5m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF30738573
Nickel-iron alloy, Ni80Fe20,
5m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF56542013
Nickel-iron alloy, Ni80Fe20,
2m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF52165773
Nickel-iron alloy, Ni80Fe20,
500mm rod, 2.0mm diameter, as drawn
Expand
View Pricing
GF42101097
Nickel-iron alloy, Ni80Fe20,
0.5m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF16331857
Nickel-iron alloy, Ni80Fe20,
5m wire, 0.5mm diameter, as drawn
Expand
View Pricing
GF07288533
Nickel-iron alloy, Ni80Fe20,
2m wire, 0.5mm diameter, as drawn
Expand
View Pricing
GF65932803
Nickel-iron alloy, Ni80Fe20,
foil, 50 x 50mm, 3.0mm thickness, as rolled
Expand
View Pricing
GF71388265
Nickel-iron alloy, Ni80Fe20,
50g lump, 5mm max. lump size
Expand
View Pricing
GF41935585
Nickel-iron alloy, Ni80Fe20,
20g lump, 5mm max. lump size
Expand
View Pricing
GF57705161
Nickel-iron alloy, Ni80Fe20,
1m wire, 0.5mm diameter, as drawn
Expand
View Pricing
Page 1 of 2
Page 1 of 2