Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nickel-iron alloy, Ni70Fe30
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
GF29100855
Nickel-iron alloy, Ni70Fe30,
100m wire, 0.03mm diameter, annealed
Expand
View Pricing
GF74320567
Nickel-iron alloy, Ni70Fe30,
50m insulated wire, 0.025mm conductor diameter, 0.0035mm insulation thickness, polyimide insulation
Expand
View Pricing
GF33864138
Nickel-iron alloy, Ni70Fe30,
20m wire, 0.03mm diameter, annealed
Expand
View Pricing
GF12567786
Nickel-iron alloy, Ni70Fe30,
100m insulated wire, 0.025mm conductor diameter, 0.0035mm insulation thickness, polyimide insulation
Expand
View Pricing
GF40183691
Nickel-iron alloy, Ni70Fe30,
10m insulated wire, 0.025mm conductor diameter, 0.0035mm insulation thickness, polyimide insulation
Expand
View Pricing
GF02895543
Nickel-iron alloy, Ni70Fe30,
50m wire, 0.03mm diameter, annealed
Expand
View Pricing
GF20028046
Nickel-iron alloy, Ni70Fe30,
20m insulated wire, 0.025mm conductor diameter, 0.0035mm insulation thickness, polyimide insulation
Expand
View Pricing
GF55464712
Nickel-iron alloy, Ni70Fe30,
5m wire, 0.03mm diameter, annealed
Expand
View Pricing
GF48425519
Nickel-iron alloy, Ni70Fe30,
10m wire, 0.03mm diameter, annealed
Expand
View Pricing
GF59168166
Nickel-iron alloy, Ni70Fe30,
5m insulated wire, 0.025mm conductor diameter, 0.0035mm insulation thickness, polyimide insulation
Expand
View Pricing