Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nickel(II) oxide
Linear Formula:
NiO
CAS Number:
1313-99-1
Molecular Weight:
74.69
Products
Product Category
anode materials (2)
Brand
Sigma-Aldrich (6)
Sustainability
Color
Feature
Formula Weight
Physical Form
Purity
Chemical Composition
Quality Segment
Application
Particle Size (µm)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
203882
Nickel(II) oxide,
99.99% trace metals basis
Expand
View Pricing
637130
Nickel(II) oxide,
nanopowder, <50 nm particle size (TEM), 99.8% trace metals basis
Expand
View Pricing
399523
Nickel(II) oxide,
green, −325 mesh, 99%
Expand
View Pricing
481793
Nickel(II) oxide,
≥99.995% trace metals basis
Expand
View Pricing
774545
Nickel(II) oxide,
nanowires, diam. × L ~20 nm × 10 μm
Expand
View Pricing
1.00782
Nickel(II) oxide,
Technipur
®
Expand
View Pricing